Learn More
MilliporeSigma™ Calbiochem™ Bleocin Streptomyces verticillus
A unique antibiotic of the bleomycin family that is toxic to eukaryotic and prokaryotic cells.
Manufacturer:  MilliporeSigma™ 20340810MG
 View more versions of this product
Catalog No. 20-340-810MG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
BLEOCIN™ Antibiotic cleaves double-stranded DNA and inhibits DNA synthesis, resulting in cell death. Can be used for selection of recombinant clones in a variety of cell types transfected with vectors containing a
Specifications
Specifications
≥85% by HPLC
2-8°C
10mg