Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bleocin Streptomyces verticillus

A unique antibiotic of the bleomycin family that is toxic to eukaryotic and prokaryotic cells.

Specifications

Storage Requirements 2-8°C
Purity ≥85% by HPLC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
20-340-810MG
View Documents
MilliporeSigma™
20340810MG
10mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
20-340-8250MG
View Documents
MilliporeSigma™
203408250MG
250mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
20-340-8100MG
View Documents
MilliporeSigma™
203408100MG
100mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

BLEOCIN™ Antibiotic cleaves double-stranded DNA and inhibits DNA synthesis, resulting in cell death. Can be used for selection of recombinant clones in a variety of cell types transfected with vectors containing a
Specifications

Specifications

2-8°C
≥85% by HPLC
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit