Learn More
MilliporeSigma™ Calbiochem™ BMP-2 Recombinant Human Protein
expressed in E. coli and purified by proprietary chromatographic methods
Supplier:  MilliporeSigma™ 2036412UG
Catalog No. 20-364-12UG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Recombinant, human bone morphogenetic protein-2 (BMP-2) expressed in E. coli and purified by proprietary chromatographic methods. BMP-2 is a homodimeric protein that belongs to the transforming growth factor-β (TGF-β) superfamily. BMP-2 is known to regulate diverse fundamental processes during embryonic development. It also plays an important role in cardiac morphogenesis and is expressed in a variety of tissues such as lung, spleen, brain, liver, prostate, ovary, and small intestine.
Specifications
Specifications
Lyophilized from sterile 10mM sodium citrate, pH 3.5
2 μg
E. coli
-20°C