Learn More
MilliporeSigma™ Calbiochem™ Carrier-Free Mitomycin C
Antibiotic and carcinostatic agent.
Manufacturer:  MilliporeSigma™ 47582010MG
Catalog No. 47-582-010MG
Add to cart
Description
Description
Inhibits DNA synthesis by cross-linking DNA at guanine and adenine residues; disrupts base pairing. Forms covalent adducts into deoxyguanine and guanine under acidic conditions. Induces apoptosis in gastric cancer cells. Blocks the cell cycle at the G
Specifications
Specifications
10mg
≥95% by HPLC
2-8°C