Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cefotaxime Sodium salt High Purity

Potent β-lactamase-resistant antibiotic of cephalosporin class.

$182.00

Specifications

Storage Requirements 15-30°C
Purity ≥97% by HPLC
Antibiotic Spectrum Active against Gram-positive, Gram-negative organisms, including Gram-negative anaerobes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
21-950-41GM
View Documents
MilliporeSigma™
2195041GM
1g
Each for $182.00
Add to cart
 
21-950-45GM
View Documents
MilliporeSigma™
2195045GM
5g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Active against Gram-positive and Gram-negative organisms, including Gram-negative anaerobes. Inhibits cell wall synthesis.
Specifications

Specifications

15-30°C
≥97% by HPLC
Active against Gram-positive, Gram-negative organisms, including Gram-negative anaerobes
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit