Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cell Culture Tested G 418 Sulfate

Sterile-filtered solution of Cat. No. Sterile-Filtered Aqueous Solution, Cell Culture Tested - CAS 108321-42-2Sterile-Filtered Aqueous Solution, Cell Culture Tested - CAS 108321-42-2

Specifications

Storage Requirements 2-8°C
Purity ≥98% by TLC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
34-581-210ML
View Documents
MilliporeSigma™
34581210ML
10mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
34-581-220ML
View Documents
MilliporeSigma™
34581220ML
20mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
34-581-250ML
View Documents
MilliporeSigma™
34581250ML
50mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Sterile-filtered solution of Cat. No. Sterile-Filtered Aqueous Solution, Cell Culture Tested - CAS 108321-42-2Sterile-Filtered Aqueous Solution, Cell Culture Tested - CAS 108321-42-2
Specifications

Specifications

2-8°C
≥98% by TLC
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit