Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cell Culture Tested G 418 Sulfate

an aminoglycoside antibiotic related to Gentamicin

$81.50 - $4200.00

Specifications

Storage Requirements 15-30°C
Purity ≥98% by TLC
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
34-581-0100GM
View Documents
MilliporeSigma™
345810100GM
100gm
Each for $4,200.00
Add to cart
 
34-581-0250MG
View Documents
MilliporeSigma™
345810250MG
250mg
Each for $81.50
Add to cart
 
34-581-025GM
View Documents
MilliporeSigma™
34581025GM
25g
Each for $2,050.00
Add to cart
 
34-581-0500MG
View Documents
MilliporeSigma™
345810500MG
500mg
Each for $110.10
Add to cart
 
34-581-01GM
View Documents
MilliporeSigma™
3458101GM
1g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
34-581-05GM
View Documents
MilliporeSigma™
3458105GM
5g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

G418 also known as Geneticin™ is an aminoglycoside antibiotic related to Gentamicin. Used as a selective agent in transfection of eukaryotic cells. Has highest potency ≥730μg/mg and purity ≥98%.
Specifications

Specifications

15-30°C
≥98% by TLC
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit