Learn More
MilliporeSigma™ Calbiochem™ Human Recombinant TGF-beta1
Recombinant, human transforming growth factor-β1 expressed in CHO cells.
Manufacturer:  MilliporeSigma™ 6164552UG
Catalog No. 61-645-52UG
Add to cart
Description
Description
Promotes apoptosis in resting human B lymphocytes, glioma cells, and trigeminal neurinomal cells. Induces the accumulation of extracellular matrix in various diseases.
Specifications
Specifications
-20°C
Lyophilized from a sterile filtered solution in 50μg BSA/μg TGF-beta1, 35% acetonitrile, 0.1% TFA
Induces accumulation of extracellular matrix in various diseases
2μg
CHO cells