Learn More
MilliporeSigma™ Calbiochem™ γ-Irradiated Neomycin Sulfate
Sterilized by γ-irradiation. An aminoglycoside antibiotic that inhibits translation by binding to the 30S ribosomal subunit. Potency (anhydrous): ≥600µg/mg.
Manufacturer:  MilliporeSigma™ 48010020ML
Catalog No. 48-010-020ML
Add to cart
Description
Description
Specifications
Specifications
20mL
-20° C