Learn More
MilliporeSigma™ Calbiochem™ Kanamycin Sulfate Available on GSA/VA Contract for Federal Government customers only.
An aminoglycoside antibiotic effective against Gram-positive and Gram-negative organisms.
$82.50 - $1173.00
Specifications
Storage Requirements 15-30°C
Antibiotic Spectrum Effective against Gram-positive and Gram-negative organisms
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
42-031-15GM Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
4203115GM
5g
Each for $82.50
Add to cart
42-031-125GM
View Documents
MilliporeSigma™
42031125GM
25g
Each for $361.50
Add to cart
42-031-1500GM
View Documents
MilliporeSigma™
420311500GM
500g
Each for $1,173.00
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
15-30°C
Effective against Gram-positive and Gram-negative organisms