Learn More
MilliporeSigma™ Calbiochem™ Mitomycin C
Antibiotic and carcinostatic agent.
Manufacturer:  MilliporeSigma™ 475892MG
Catalog No. 47-589-2MG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Inhibits DNA synthesis by cross-linking DNA at guanine and adenine residues; disrupts base pairing. Forms covalent adducts into deoxyguanine and guanine under acidic conditions. Induces apoptosis in gastric cancer cells. Blocks the cell cycle at the G
Specifications
Specifications
2mg
≥95% by HPLC
2-8°C