Learn More
MilliporeSigma™ Calbiochem™ Neomycin Sulfate
an aminoglycoside antibiotic that inhibits translation by binding to the small subunit of prokaryotic ribosomes.
Manufacturer:  MilliporeSigma™ 480125GM
Catalog No. 48-012-5GM
Add to cart
Description
Description
Recently shown to inhibit angiogenin-induced angiogenesis. Blocks voltage-sensitive Ca
Specifications
Specifications
25g
15-30°C