Learn More
EMD Millipore Calbiochem™ OmniPur™ G418 Sulfate
G418 is an aminoglycoside antibiotic related to gentamycin that inhibits prokaryotic and eukaryotic protein synthesis.
Manufacturer:  EMD Millipore 5.09290.0002
 View more versions of this product
Catalog No. 50-929-00002
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
4 to 6
≥90% by HPLC
1g
15-30°C