Learn More
EMD Millipore Calbiochem™ OmniPur™ G418 Sulfate
G418 is an aminoglycoside antibiotic related to gentamycin that inhibits prokaryotic and eukaryotic protein synthesis.
Manufacturer:  EMD Millipore 5.09290.0003
 View more versions of this product
Catalog No. 50-929-00003
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Aminoglycoside related to gentamycin that inhibits prokaryotic and eukaryotic protein synthesis. Toxic to bacteria, yeast, protozoans, helminths, higher plant, and mammalian cells. Widely used in the selection of eukaryotic expression vectors carrying the bacterial
Description & Specifications
Specifications
4 to 6
≥90% by HPLC
5g
15-30°C