Learn More
MilliporeSigma™ Calbiochem™ OmniPur™ G418 Sulfate
G418 is an aminoglycoside antibiotic related to gentamycin that inhibits prokaryotic and eukaryotic protein synthesis.
Specifications
Purity ≥90% by HPLC
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-929-00005
View Documents
MilliporeSigma™
5.09290.0005
100gm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Aminoglycoside related to gentamycin that inhibits prokaryotic and eukaryotic protein synthesis. Toxic to bacteria, yeast, protozoans, helminths, higher plant, and mammalian cells. Widely used in the selection of eukaryotic expression vectors carrying the bacterial
Specifications
Specifications
≥90% by HPLC