Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Oxytetracycline, Hydrochloride

An antimicrobial agent

$198.80

Specifications

Storage Requirements 2-8°C
Purity ≥95% by HPLC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-010-5100GM
View Documents
MilliporeSigma™
500105100GM
100gm
Each for $198.80
Add to cart
 
50-010-510GM
View Documents
MilliporeSigma™
50010510GM
10g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

A member of the tetracycline family that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit.
Specifications

Specifications

2-8°C
≥95% by HPLC
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit