Learn More
MilliporeSigma™ Calbiochem™ Penicillin G, Potassium Salt
Antibiotic effective against Gram-positive bacteria. Blocks cell wall formation in growing cells, resulting in accumulation of uracil nucleotides of muramic acid peptides.
Specifications
Storage Requirements 2-8°C
Antibiotic Spectrum Effective against Gram-positive bacteria
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
51-612-5MU
View Documents
MilliporeSigma™
516125MU
25MU This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
51-611-00MU
View Documents
MilliporeSigma™
5161100MU
100MU This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Specifications
Specifications
2-8°C
Effective against Gram-positive bacteria