Learn More
EMD Millipore™ Calbiochem™ Pimaricin, Streptomyces chattanoogensis Available on GSA/VA Contract for Federal Government customers only.
Antifungal polyene macrolide that is active in the cell membrane.
Manufacturer:  Millipore Sigma 527962100MG
 View more versions of this product
Catalog No. 52-796-2100MG
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
≥80% by HPLC
100mg
15-30°C