Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Pimaricin, Streptomyces chattanoogensis Available on GSA/VA Contract for Federal Government customers only.

Antifungal polyene macrolide that is active in the cell membrane.

$269.50 - $1470.00

Specifications

Storage Requirements 15-30°C
Purity ≥80% by HPLC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
52-796-2100MG Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
527962100MG
100mg
Each for $269.50
Add to cart
 
52-796-21GM
View Documents
MilliporeSigma™
5279621GM
1g
Each for $1,470.00
Add to cart
 
Description

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

15-30°C
≥80% by HPLC
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit