Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protein A, Biotin Conjugate

Catalog No. 2031952mg Shop All MilliporeSigma Products
Click to view available options
Quantity:
2 mg

Native protein A conjugated to biotin

  • Recognizes the Fc portions of immunoglobulins of various species
  • Form: Liquid
  • Formulation: In 10mM PBS, 1% BSA, pH 7.6
  • Purity: Single band at M.W. 50,000 by SDS-PAGE
  • Preservative: ≤0.1% sodium azide
TRUSTED_SUSTAINABILITY

Specifications

Description Protein A, Biotin Conjugate
Quantity 2 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.