Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Recombinant Brain Derived Neurotrophic Factor

expressed in E. coli

$334.00 - $524.00

Specifications

Storage Requirements -20°C
Source E. coli
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
20-370-25UG
View Documents
MilliporeSigma™
2037025UG
5μg
Each for $334.00
Add to cart
 
20-370-210UG
View Documents
MilliporeSigma™
20370210UG
10μg
Each for $524.00
Add to cart
 
Description

Description

Recombinant, human brain drived neurotrophic factor expressed in E. coli. Promotes survival and growth of neurons. Structurally and functionally related to nerve growth factor. Its actions are mediated via the TrkB receptor.
Specifications

Specifications

-20°C
E. coli
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit