Learn More
MilliporeSigma™ Calbiochem™ Recombinant Human beta-Nerve Growth Factor
Recombinant, human β-nerve growth factor expressed in NSO cells. Involved in neuroimmune interactions and inflammation.
Manufacturer:  MilliporeSigma™ 480275100UG
Catalog No. 48-027-5100UG
Add to cart
Description
Description
Specifications
Specifications
100μL
Lyophilized from sterile-filtered solution in PBS containing 50μg BSA/μg beta-NGF
NSO cells
≤-70°C