Learn More
MilliporeSigma™ Calbiochem™ Recombinant Human FGF-10
consisting of amino acids 137-208 and expressed in E. Coli.
Manufacturer:  MilliporeSigma™ 34159925UG
Catalog No. 34-159-925UG
Add to cart
Description
Description
Recombinant, human fibroblast growth factor 10 (FGF-10) consisting of amino acids 137-208 and expressed in E. Coli. Mature human FGF-10, a ∽20 kDa protein, contains a serine-rich region near its N-terminus sharing 93% amino acid sequence identity with mouse FGF-10 and 96% with rat FGF-10.
Specifications
Specifications
-20°C
25μg
Lyophilized from 0.2μm sterile-filtered MOPS, Na2SO4, EDTA, DTT, BSA.
E. coli