Learn More
MilliporeSigma™ Calbiochem™ Sterile Nystatin
Antifungal antibiotic effective against yeast and mycoplasma.
Manufacturer:  MilliporeSigma™ 47592150MG
Catalog No. 47-592-150MG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Sterilized by γ-irradiation. Antifungal antibiotic effective against yeast and mycoplasma. The complex contains 3 biologically active components, A1, A2, and A3. Reconstitute in 10mL sterile deionized water to prepare a 5mg/mL solution.
Specifications
Specifications
50mg
-20° C