Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Streptavidin

High-affinity biotin binding protein

$120.10 - $618.50

Specifications

Description Streptavidin
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-973-010MG
View Documents
MilliporeSigma™
18973010MG
10mg
Each for $618.50
Add to cart
 
18-973-01MG
View Documents
MilliporeSigma™
1897301MG
1mg
Each for $120.10
Add to cart
 
Description

Description

  • Form: Lyophilized solid
  • Molar Mass; 60,000
  • Specific Activity: ≥10 units/mg protein
  • Synonyms: Avidin, Streptomyces avidinii
  • Soluble in water or many low ionic strength buffers at neutral pH
  • Storage: Store at -20°C and protect from moisture
Specifications

Specifications

Streptavidin
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit