Learn More
EMD Millipore™ Calbiochem™ Streptozotocin Available on GSA/VA Contract for Federal Government customers only.
An N-nitroso-containing diabetogenic compound that acts as a nitric oxide donor in pancreatic islets
Manufacturer:  Millipore Sigma 5722011GM
 View more versions of this product
Catalog No. 57-220-11GM
  Check Availability
Add to cart
Specifications
Description
An antibiotic effective against Gram-negative bacteria. Causes DNA alkylation and DNA strand breaks in pancreatic islet cells.
Description & Specifications
Specifications
Effective against Gram-negative bacteria
≥95% by HPLC
1g
-20° C