EMD Millipore™ Calmodulin, Bovine Brain, Protein

Manufacturer: EMD Millipore 2086905MG

 View more versions of this product

Catalog No. 20-869-05MG

  Check Availability
Add to Cart

    Specifications

    Preparation Method Aqueous buffers or H2O
    Purification Method SDS-PAGE
    Purity ≥95%
    Quantity 5mg
    Recombinant No
    Immunogen Calmodulin
    Storage Requirements Following reconstitution, aliquot and freeze (−20°C or −70°C). Stock solutions are stable for up to 2 months at −20°C or for up to 4 months at −70°C.
    Regulatory Status RUO
    Format Lyophilized
    Formulation Lyophilized from 150mM NaCl, 50mM Tris-HCl, 2mM EDTA, pH 7.6.
    Host Species Bovine brain

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit