EMD Millipore™ Cathepsin D, Human Liver, Protein

Manufacturer: EMD Millipore 21940130U

 View more versions of this product

Catalog No. 21-940-130U

  Check Availability
Add to Cart

    Specifications

    Purification Method SDS-PAGE
    Preparation Method Sterile H2O or aqueous buffers. Please see vial label for lot-specific reconstitution volume.
    Purity ≥90%
    Quantity 30U
    Recombinant No
    Immunogen Cathepsin D
    Storage Requirements Following reconstitution, aliquot and freeze (−20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 1 week at 4°C. Keep on ice when working with material. When product is brought to its active pH (3.3), autolysis occurs.
    Regulatory Status RUO
    Format Lyophilized
    Formulation Lyophilized from 2mM sodium phosphate buffer, pH 6.5.
    Host Species Human liver

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit