Learn More
EMD Millipore Centriplus™ Centrifugal Concentrators†
Provide fast, convenient, high-recovery alternatives to dialysis and ethanol precipitation
Manufacturer:  EMD Millipore 42413/BAD PRICING
 View more versions of this product
Catalog No. 42413
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Disclaimers:
An Amicon Bioseparations product.
Specifications Description & Specifications
Specifications
Microcon
Centricon YM-100
1 in.
Polypropylene
4.33 in.
3,000xG
2.34cm2
Regenerated Cellulose
Blue
29mm
Sample reservoir; Base; Retentate vial; Caps
110mm
100,000 d
0.5mL