EMD Millipore Centriprep™ Centrifugal Filter Units Available on GSA/VA Contract for Federal Government customers only.
For processing high-solute samples
Manufacturer: EMD Millipore 4311
 View more versions of this product
Catalog No. 4311
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Length (Metric) 135mm
    MWCO 50,000 d
    Quantity 96/Pk.
    Surface Area 2.84cm2
    Volume (Metric) 12 to 15mL
    Membrane Material Ultracel-YM Regenerated Cellulose
  • Description & Specifications
Related Products
Product Content Feedback