Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Centriprep™ Centrifugal Filter Units
For processing high-solute samples
Supplier: MilliporeSigma™ 4310
Description
For processing high-solute samples
Centriprep™ centrifugal filters are disposable ultrafiltration devices used for purifying, concentrating, and desalting biological samples in the 2–15 mL volume range. These complete, ready-to-use ultrafiltration devices are designed for operation in most centrifuges that can accommodate 50 mL centrifuge tubes. They are easy to use and offer a high flow rate. Centriprep™ devices consist of a sample container with a twist-lock cap, a filtrate collector containing a low-adsorptive, Ultracel™ regenerated cellulose membrane and an air-seal cap for sample isolation.Features & Benefits:
Applications:Concentration and Purification of Particle-laden Solutions, Solutions with High Solute Concentration, Separation of Low MW Solutes from Fermentation Broths, Cell Culture Media and Lysates
Specifications
Single-use | |
50 kDa | |
2.84 cm2 | |
24Pack | |
12 to 15 mL |
Ultracel-YM Regenerated Cellulose | |
Non-sterile | |
Regenerated Cellulose (RC) | |
Centrifugal Filters | |
135 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction