Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Centriprep™ Centrifugal Filter Units Available on GSA/VA Contract for Federal Government customers only.

For processing high-solute samples

$1528.00

Specifications

Material (Membrane) Ultracel-YM Regenerated Cellulose
Length (Metric) 135 mm
Filtration Area 2.84 cm2
Volume (Metric) 12 to 15 mL
View More Specs

Products 7
Catalog Number Mfr. No. MWCO Quantity Price Quantity & Availability  
Catalog Number Mfr. No. MWCO Quantity Price Quantity & Availability  
4311
View Documents
MilliporeSigma™
4311
50,000 d 96Pack
Each for $1,528.00
Add to cart
 
4304 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
4304
10,000 d 24Pack This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
4305
View Documents
MilliporeSigma™
4305
10,000 d 96Pack This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
4302 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
4302
3,000 d 24Pack This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
4307
View Documents
MilliporeSigma™
4307
30,000 d 96Pack This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
4306
View Documents
MilliporeSigma™
4306
30,000 d 24Pack This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
4310
View Documents
MilliporeSigma™
4310
50,000 d 24Pack This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

For processing high-solute samples

Centriprep™ centrifugal filters are disposable ultrafiltration devices used for purifying, concentrating, and desalting biological samples in the 2–15 mL volume range. These complete, ready-to-use ultrafiltration devices are designed for operation in most centrifuges that can accommodate 50 mL centrifuge tubes. They are easy to use and offer a high flow rate. Centriprep™ devices consist of a sample container with a twist-lock cap, a filtrate collector containing a low-adsorptive, Ultracel™ regenerated cellulose membrane and an air-seal cap for sample isolation.

Features & Benefits:

  • For use with samples containing high levels of solutes
  • Unique inverse flow mode of operation with large deadstop
  • Fast sample processing
  • Available with Ultracel™ membranes: (3, 10, 30 and 50 kDa NMWL)
  • Fits standard swinging-bucket rotor for 50 mL tubes
  • Applications:Concentration and Purification of Particle-laden Solutions, Solutions with High Solute Concentration, Separation of Low MW Solutes from Fermentation Broths, Cell Culture Media and Lysates

    Specifications

    Specifications

    Ultracel-YM Regenerated Cellulose
    135 mm
    2.84 cm2
    12 to 15 mL
    SDS
    Product Certifications
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit