EMD Millipore Centriprep™ Centrifugal Filter Units Available on GSA/VA Contract for Federal Government customers only.
For processing high-solute samples
Manufacturer: EMD Millipore 4303
 View more versions of this product
Catalog No. 4303
  Check Availability
Add to Cart
    Specifications
    Length (Metric) 135mm
    MWCO 3,000 d
    Quantity 96/Pk.
    Surface Area 2.84cm2
    Volume (Metric) 12 to 15mL
    Membrane Material Ultracel-YM Regenerated Cellulose
  • Description & Specifications
Related Products
Product Content Feedback