Learn More
EMD Millipore Centriprep™ Centrifugal Filter Units Available on GSA/VA Contract for Federal Government customers only.
For processing high-solute samples
Manufacturer:  EMD Millipore 4304
 View more versions of this product
Catalog No. 4304
  Check Availability
Add to cart
Specifications
Description
  • Fast sample processing
  • High-recovery, Ultracel™-YM membrane
  • Ideal for concentrating and purifying particle-laden solutions or high concentrations
  • Separate low MW solutes from fermentation broths, cell culture media, and cell lysates
  • Unique inverse flow mode with large deadstop
  • Use in 50mL tube swinging-bucket or fixed-angle rotor
Description & Specifications
Specifications
Ultracel-YM Regenerated Cellulose
135mm
24/Pk.
12 to 15mL
Green
10,000 d
2.84cm2
Promotions
Promotions
  • Millipore Sigma
    When you spend $1,000 on EMD Millipore™ Stericup™, Steritop™, Steriflip™, Millex™, Amicon Ultra™, Immobilon™ and Luminata™ products listed below, you’ll get an additional $1,000 to spend on EMD Millipore products. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.
    06-30-2018