Learn More
MilliporeSigma™ Certistain™ Pyronine G Dye
Pyronine G (C.I. 45005) for microscopy Certistain™. CAS 92-32-0, molar mass 302.81g/mol.
Supplier:  MilliporeSigma™ 1.07518.0025
Catalog No. M1075180025
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Pyronine G (C.I. 45005) is a Certistain™ dry dye for DNA and RNA staining in methyl green-pyronine G staining in histological sections. It is also described as a dye for staining RNA and DNA.
Specifications
Specifications
25g
Pyronine G Dye
Stains DNA and RNA in methyl green-pyronine G staining in histological sections
250-260°C