Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Chemicon Recombinant Rat Ciliary Neurotrophic Factor
Potent neural factor that was characterized as survivability factor for chick ciliary neurons in vitro
Manufacturer: MilliporeSigma GF035
Description
Recommended Storage
Store dessicated at 0°C
Specifications
22.7kd | |
25µg | |
Store dessicated at 0°C | |
Rat |
>98% by SDS-PAGE and HPLC analysis | |
E.coli | |
ED50 as determined by dose-dependent proliferation of human TF-1 cells is ≤15-30ng/mL | |
Sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title