EMD Millipore™ Chorionic Gonadotropin, Human Urine, Standard Grade, Protein Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: EMD Millipore 2307341MG

 View more versions of this product

Catalog No. 23-073-41MG

  Check Availability
Add to Cart

    Specifications

    Preparation Method H2O
    Purification Method By weight
    Purity ≥30%, Standard Grade
    Quantity 1mg
    Recombinant No
    Immunogen Chorionic Gonadotropin
    Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Avoid freeze/thaw cycles of reconstituted solutions. Stock solutions are stable for up to 3 months at −20°C.
    Regulatory Status RUO
    Format Off-white solid
    Host Species Human urine

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit