EMD Millipore™ Collagenase, Type I, Clostridium histolyticum, Protein Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: EMD Millipore 2341531GM

 View more versions of this product

Catalog No. 23-415-31GM

  Check Availability
Add to Cart

    Specifications

    Preparation Method Aqueous Buffers
    Quantity 1g
    Recombinant No
    Source Clostridium histolyticum
    Immunogen Collagenase, Type I
    Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 2 months at −20°C.
    Regulatory Status RUO
    Format Lyophilized

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit