Learn More
MilliporeSigma colorpHast pH Strips
Accurate results are achieved even in weakly buffered, turbid and colored samples. Each pack contains 100 strips.
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
M95903
|
MilliporeSigma
95903 |
N/A | Range: Full, pH 0 to 14 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Unique pH test papers feature covalently bound indicators that will not bleed, dilute, or contaminate trace samples
- Strips give sharp, clear color changes for easy comparison to color scale provided on the colorpHast package Full-range (Universal) Strips measure 0 to 14pH range in a single test with 0.5pH sensitivityeach strip includes four squares with different indicators Intermediate-range Strips with three squares each have a sensitivity of 0.3 to 0.5 Narrow-range Strips with one pad have a sensitivity of 0.2 to 0.3
Order Info
Each pack contains 100 strips