EMD Millipore™ Concanavalin A, Canavalia ensiformis, Protein Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: EMD Millipore 2345671GM

 View more versions of this product

Catalog No. 23-456-71GM

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Following reconstitution, aliquot and freeze (−20°C) or add 0.08% NaN3 and refrigerate (4°C). Stock solutions are stable for up to 6 months at −20°C or for up to 3 months at 4°C.
    Format Lyophilized
    Preparation Method To dissolve, add Con A slowly to a solution of 10mM HEPES buffered saline, 100μM CaCl2, pH 8.5. Allow the lectin to dissolve without agitation.
    Quantity 1g
    Recombinant No
    Source Canavalia ensiformis
    Immunogen Concanavalin A
    Regulatory Status RUO
    Formulation Salt-free.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit