EMD Millipore™ Cytochrome c, Equine Heart, Protein

Manufacturer: EMD Millipore 250600100MG

 View more versions of this product

Catalog No. 25-060-0100MG

  Check Availability
Add to Cart

    Specifications

    Preparation Method H2O (100mg/mL)
    Purification Method A550
    Purity ≥90%
    Quantity 100mg
    Recombinant No
    Immunogen Cytochrome c
    Storage Requirements Following reconstitution, refrigerate (4deg;C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 2 weeks at 4°C, for up to 6 months at −20°C, and for up to 3 days at room temperature.
    Regulatory Status RUO
    Format Salt-free lyophilized solid
    Host Species Equine heart

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit