Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Dulbecco's Modified Eagle Media 1X, Liquid, w/4,500mg/L Glucose

Optimized and validated for cell culture.

$23.25 - $40.85

Specifications

Storage Requirements 2-8°C for 10 months
For Use With (Application) Cell culture
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SLM020A
View Documents
MilliporeSigma™
SLM020A
1L
Each for $40.85
Add to cart
 
SLM020B
View Documents
MilliporeSigma™
SLM020B
500mL
Each for $23.25
Add to cart
 
Description

Description

Specifications

Specifications

2-8°C for 10 months
Cell culture
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit