Learn More
MilliporeSigma Durapore PVDF Membrane Filters: Hydrophilic: 0.10µ Pore Size
Recommended when maximum recovery is important
$120.60 - $254.00
Specifications
Autoclavable | Yes |
---|---|
Bubble Point | 70psi |
Color | White |
Material | Polyvinylidene Fluoride (PVDF) |
Sterility | Sterile |
Catalog Number | Mfr. No. | Diameter (Metric) Outer | Quantity (No. per Case) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Outer | Quantity (No. per Case) | Price | Quantity & Availability | ||||
VVLP01300
|
MilliporeSigma
VVLP01300 |
13mm | 100/pk. |
Pack of 100 for $120.60
|
|
VVLP04700
![]() |
MilliporeSigma
VVLP04700 |
47mm | 100/pk. |
Pack of 100 for $185.30
|
|
VVLP02500
![]() |
MilliporeSigma
VVLP02500 |
25mm | 100/pk. |
Pack of 100 for $143.10
|
|
VVLP09050
|
MilliporeSigma
VVLP09050 |
90mm | 50/pk. |
Pack of 50 for $254.00
|
|
Description
Recommended when maximum recovery is important
- Mean thickness: 125µm
- Typical refractive index: 1.42
- Autoclavable
- Hydrophilic form is made from modified polyvinylidene fluoride (PVDF); lower protein binding than nitrocellulose, nylon, or PTFE
Applications:
- Clarification of air and other gases
- Sterilizing filtration of protein solutions, tissue culture media, additives, antibiotics, and alcohols
- Particulate removal; biological assays (receptor binding studies, TCA precipitates)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Yes | |
White | |
Sterile |
70psi | |
Polyvinylidene Fluoride (PVDF) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.