Learn More
MilliporeSigma™ Durapore™ PVDF Membrane Filters: 0.22μ Pore Size
Recommended when maximum recovery is important. Durapore™ PVDF Membranes: 0.22μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
Manufacturer: MilliporeSigma™ GVWP14250
Description
Recommended when maximum recovery is important. Durapore™ PVDF Membranes: 0.22μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
- Composed of polyvinylidene fluoride (PVDF) that binds far less protein than than nylon, nitrocellulose or PTFE membranes
- White, plain membranes are autoclavable
- Hydrophilic membranes are 125µm thick with 0.22µm pores
- Typical refractive index: 1.42
- Clarification of air and other gases
- Sterilizing filtration of protein solutions, tissue culture media, additives, antibiotics and alcohols
- Particulate removal and biological assays (receptor binding studies, TCA precipitates)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Yes | |
White | |
2L/min/cm2 | |
Polyvinylidene Fluoride (PVDF) | |
50/pk. | |
Hydrophilic |
50psi | |
142mm | |
6.9mL/min./cm2 | |
0.22µm | |
125µm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.