Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EmbryoMax™ 1X Dulbecco's Phosphate Buffered Saline w/o Ca++ and Mg++

Used for routine mouse embryonic stem cell culture applications.

$34.25

Specifications

Storage Requirements 15-30°C
For Use With (Application) Stem cell culture
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BSS1006A
View Documents
MilliporeSigma™
BSS1006A
1L
Each for $34.25
Add to cart
 
BSS1006B
View Documents
MilliporeSigma™
BSS1006B
500mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

The EmbryoMax 1X Dulbecco's Phosphate Buffered Saline w/o Ca++ and Mg++ is available in a 500mL format and may be used for routine mouse embryonic stem cell culture applications.
Specifications

Specifications

15-30°C
Stem cell culture
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit