Learn More
MilliporeSigma™ EmbryoMax™ KSOM Embryo Culture (1x) w/o Phenol Red
Optimized and validated for Embryo Culture.
Manufacturer:  MilliporeSigma™ MR020P5F
 View more versions of this product
Catalog No. MR020P5F
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
We now offer our most popular embryo culture media in a dry powder format. Each has been formulated for optimal, consistent performance. This new dry powdered format offers greatly increased shelf-life, easy and quick preparation, and the freshest nutrients to support the development of your embryos in culture. Just add the sterile diluent (included in the kit) to the powder, mix gently, and filter to prepare FRESH medium with the correct pH and osmolarity.
Specifications
Specifications
2-8°C
5 x 10 mL