Learn More
MilliporeSigma™ EmbryoMax™ M2 Medium (1X) Powdered With Phenol Red
Optimized and validated for Embryo Culture.
Manufacturer:  MilliporeSigma™ MR015P5F
 View more versions of this product
Catalog No. MR015P5F
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Dry Powdered form offers easy and quick preparation, and the freshest nutrients to support the development of embryos in culture. Just add the sterile diluent (included in the kit) to the powder, mix gently, and filter to prepare FRESH medium with the correct pH and osmolarity.
Specifications
Specifications
Stem cell culture and embryo culture
2- 8°C
50mL