Learn More
MilliporeSigma™ EmbryoMax™ M2 Medium (1x)
Optimized and validated for Embryo Culture.
$39.75
Specifications
For Use With (Application) Embryo and stem cell culture
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MR051F
View Documents
MilliporeSigma™
MR051F
10mL
Each for $39.75
Add to cart
MR015PD
View Documents
MilliporeSigma™
MR015PD
50mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Dry Powdered form offers easy and quick preparation, and the freshest nutrients to support the development of embryos in culture. Just add the sterile diluent (included in the kit) to the powder, mix gently, and filter to prepare FRESH medium with the correct pH and osmolarity.
Specifications
Specifications
Embryo and stem cell culture