Learn More
MilliporeSigma™ EmbryoMax™ Modified M16 Medium (1X), Powdered, w/o phenol red
Optimized and validated for Embryo Culture.
Manufacturer:  MilliporeSigma™ MR010PD
 View more versions of this product
Catalog No. MR010PD
Add to cart
Description
Description
We now offer our most popular embryo culture media in a dry powder format. Each has been formulated for optimal, consistent performance. This new dry powdered format offers greatly increased shelf-life, easy and quick preparation, and the freshest nutrients to support the development of your embryos in culture. Just add the sterile diluent (included in the kit) to the powder, mix gently, and filter to prepare FRESH medium with the correct pH and osmolarity.
Specifications
Specifications
1 x 50mL
2- 8°C for 6 - 12 months
50mL
Stem cell culture and embryo culture