Learn More
MilliporeSigma™ EmbryoMax™ Modified M16 Medium (1X), Powdered, w/o phenol red
Optimized and validated for Embryo Culture.
$64.60 - $275.50
Specifications
For Use With (Application) Stem cell culture and embryo culture
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
MR010PD
View Documents
MilliporeSigma™
MR010PD
50mL 1 x 50mL
Each for $64.60
Add to cart
MR010P5D
View Documents
MilliporeSigma™
MR010P5D
250mL 5 x 50mL
Each for $275.50
Add to cart
MR010P5F
View Documents
MilliporeSigma™
MR010P5F
50mL 5 x 10mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
We now offer our most popular embryo culture media in a dry powder format. Each has been formulated for optimal, consistent performance. This new dry powdered format offers greatly increased shelf-life, easy and quick preparation, and the freshest nutrients to support the development of your embryos in culture. Just add the sterile diluent (included in the kit) to the powder, mix gently, and filter to prepare FRESH medium with the correct pH and osmolarity.
Specifications
Specifications
Stem cell culture and embryo culture